
1,5-Diphenylcarbazide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:140-22-7
:
Formula:C₁₃H₁₄N₄O
MDL Number:MFCD00003013
:242.28 g/mol
Merck Index:13,03353
Boiling Pt:454 °C (1013 hPa)
Melting Pt:170…172 °C
Density:1.31 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:205-403-7
:GHS07
Frequently Bought Together