p-Cresol
Catalog # EHERC15140400
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,5 g
- Seq:3035
- Environmentally Preferable:
Specifications
About this item
Synonyms: 1-Hydroxy-4-methylbenzene, 4-Methylphenol, 4-Hydroxytoluene
:
:108.14 g/mol
Merck Index:13,02604
Boiling Pt:201.8 °C (1013 hPa)
Melting Pt:32…34 °C
Density:1.034 g/cm³ (20 °C)
Flash Pt:86 °C
Storage Temperature:Ambient
EINECS:203-398-6
UN:3455
ADR:6.1,II
:GHS06
:GHS05
CAS Number:106-44-5
Formula:C₇H₈O
MDL Number:MFCD00002376