Order Entry
Slovenia
ContactUsLinkComponent
2,5-Dimethoxytoluene
2,5-Dimethoxytoluene
Catalog # MOLE47678041-25G
Supplier:  Molekula
CAS Number:  
2,5-Dimethoxytoluene
Catalog # MOLE47678041-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Boiling Pt:218 °C (1013 hPa)
Boiling Pt:218 °C (1013 hPa)
Melting Pt:15 °C
Density:1.04 g/cm³ (20 °C)
 :152.19 g/mol
EINECS:246-343-1
 :GHS07
CAS Number:24599-58-4
Formula:C₉H₁₂O₂
MDL Number:MFCD00041901