
2-Methoxy-p-cresol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:93-51-6
:
Formula:C₈H₁₀O₂
MDL Number:MFCD00002378
:138.17 g/mol
Merck Index:13,02599
Boiling Pt:219…222 °C (1013 hPa)
Density:1.09 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:202-252-9
:GHS07
Storage Temperature:Ambient
Frequently Bought Together