
4-Chlorobenzylamine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:104-86-9
:
Formula:C₇H₈ClN
MDL Number:MFCD00008121
:141.6 g/mol
Boiling Pt:215 °C (1013 hPa)
Density:1.16 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:203-245-3
:GHS07
Frequently Bought Together