

4-Chlorobenzylamine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:104-86-9
:
Formula:C₇H₈ClN
MDL Number:MFCD00008121
:141.6 g/mol
Boiling Pt:215 °C (760 mmHg)
Density:1.16 g/cm³
Flash Pt:90 °C
Storage Temperature:Ambient
EINECS:203-245-3
UN:2922
ADR:8,III
:GHS07
:GHS05
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5576 to 1.5610 (20°C, 589 nm)
- GC:≥97.5 %
- Color scale:≤50 APHA
Specification Test Results
Frequently Bought Together