ω-Bromo-3-fluoroacetophenone, Sigma-Aldrich®
Catalog # SIAL669482-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:217.04 g/mol
:M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
CAS Number:53631-18-8
Formula:C₈H₆BrFO
MDL Number:MFCD00109756