ω-Bromo-3-fluoroacetophenone
Catalog # MOLE34359168-1G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:217.04 g/mol
Storage Temperature:Refrigerator
CAS Number:53631-18-8
Formula:C₈H₆BrFO
MDL Number:MFCD00109756