Order Entry
Slovenia
Orders LinkContactUsLinkComponent
 

ω-Bromo-3-methoxyacetophenone

Glass bottle for solids, 5 g
Catalog # MOLE33674268-5G
Supplier:   Molekula
ω-Bromo-3-methoxyacetophenone
Catalog # MOLE33674268-5G
Supplier:  Molekula
CAS Number:  5000-65-7
ω-Bromo-3-methoxyacetophenone

Glass bottle for solids, 5 g

CAS Number:  5000-65-7
Price
Inquire For Price
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  2-Bromo-3'-methoxyacetophenone, Bromomethyl m-methoxyphenyl ketone, 3-Methoxyphenacyl bromide, 2-Bromo-1-(3-methoxyphenyl)ethanone, 2-Bromo-1-(3-methoxyphenyl)ethan-1-one, α-Bromo-3-methoxyacetophenone
  • CAS Number:5000-65-7
  • Formula:C₉H₉BrO₂
  • MDL Number:MFCD00000199
  • Molecular Weight:229.07 g/mol
  • Boiling Pt:285 °C (1013 hPa)
  • Melting Pt:63…64.3 °C
  • Storage Temperature:Refrigerator
  • EINECS:225-666-1
  • UN:3261
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications