4-tert-Butyl-1-bromobenzene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:3972-65-4
:
Formula:C₁₀H₁₃Br
MDL Number:MFCD00000108
Boiling Pt:232 °C (1013 hPa)
Melting Pt:15.5 °C
Density:1.2572 g/cm³ (15 °C)
:M-Clarity™ quality level = MQ200
:213.12 g/mol
Storage Temperature:Ambient
EINECS:223-599-2
Frequently Bought Together