Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Hydroquinone

Hydroquinone

Glass bottle for solids, 1 kg
Catalog # MOLE18759645-1KG
Corrosive
Health
Irritant
Environment
Supplier:   Molekula
Hydroquinone
Catalog # MOLE18759645-1KG
Supplier:  Molekula
CAS Number:  123-31-9
Hydroquinone
Hydroquinone

Glass bottle for solids, 1 kg

CAS Number:  123-31-9
Price
145,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  1,4-Dihydroxybenzene, Quinol
  • CAS Number:123-31-9
  • Formula:C₆H₄(OH)₂
  • MDL Number:MFCD00002339
  • Molecular Weight:110.11 g/mol
  • Merck Index:13,04833
  • Boiling Pt:285 °C (1013 hPa)
  • Melting Pt:172 °C
  • Density:1.332 g/cm³ (20 °C)
  • Flash Pt:165 °C
  • Storage Temperature:Ambient
  • EINECS:204-617-8
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications