Order Entry
Slovenia
ContactUsLinkComponent
Phenetole
Phenetole
Catalog # MOLE42008230-500G
Supplier:  Molekula
CAS Number:  
Phenetole
Catalog # MOLE42008230-500G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :122.17 g/mol
Merck Index:12,07374
Boiling Pt:170 °C (1013 hPa)
Melting Pt:–30 °C
Density:0.956 g/cm³ (20 °C)
Flash Pt:57 °C
Storage Temperature:Ambient
EINECS:203-139-7
UN:3271
ADR:3,III
 :GHS02
CAS Number:103-73-1
Formula:C₆H₅OC₂H₅
MDL Number:MFCD00009090