Order Entry
Slovenia
ContactUsLinkComponent
Barium diphenylamine-4-sulphonate
Barium diphenylamine-4-sulphonate
Catalog # MOLEM73443873
Supplier:  Molekula
CAS Number:  
undefined
Barium diphenylamine-4-sulphonate
Catalog # MOLEM73443873
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Diphenylamine-4-sulphonic acid barium salt, N-Phenylsulphanilic acid barium salt
 :
 :633.89 g/mol
EINECS:228-278-0
UN:1564
 :GHS07
Storage Temperature:Ambient
CAS Number:6211-24-1
 :1300-92-1
Formula:C₂₄H₂₀BaN₂O₆S₂
MDL Number:MFCD00007497
Melting Pt:300
ADR:6.1 III