4-Iodobenzonitrile, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:3058-39-7
:
Formula:C₇H₄IN
MDL Number:MFCD00051310
:229.02 g/mol
Boiling Pt:285 °C (1013 hPa)
Melting Pt:125…128 °C
UN:3439
ADR:6.1,III
:M-Clarity™ quality level = MQ100
Frequently Bought Together