Order Entry
Slovenia
ContactUsLinkComponent
 
 
 
4-(Trifluoromethoxy)benzoyl chloride 98%
 
undefined
 
 
4-(Trifluoromethoxy)benzoyl chloride 98%

 

  • Appearance:
    Clear colorless to pink Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4710 to 1.4730 (20°C, 589 nm)
  • GC:
    ≥97.5 %

 

 

Frequently Bought Together