
4-(Trifluoromethoxy)benzaldehyde
Supplier: CHEMPUR
Certificates
About this item
CAS Number:659-28-9
:
Formula:C₈H₅F₃O₂
MDL Number:MFCD00041530
:190.12 g/mol
Boiling Pt:80 °C (20 hPa)
Density:1,331 g/cm³ (20 °C)
EINECS:211-531-4
:GHS07
Storage Temperature:Ambient
Frequently Bought Together