Order Entry
Slovenia
ContactUsLinkComponent
Methyl-3-nitrobenzoate 98%
Methyl-3-nitrobenzoate 98%
Certificates
undefined
Methyl-3-nitrobenzoate 98%

Specification Test Results

  • Appearance:
    Beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    76 to 80 °C
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together