3,5-Dihydroxybenzaldehyde, Sigma-Aldrich®
Catalog # SIAL368113-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:138.12 g/mol
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:247-479-4
CAS Number:26153-38-8
Formula:C₇H₆O₃
MDL Number:MFCD00016611