
4-Bromobenzenesulphonyl chloride
Catalog # MOLE12638559-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:255.52 g/mol
Merck Index:13,01391
Boiling Pt:287 °C (1013 hPa)
Melting Pt:74 °C
Density:1.37 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-683-2
UN:3261
ADR:8,II
:GHS05
CAS Number:98-58-8
Formula:C₆H₄BrClO₂S
MDL Number:MFCD00007437