2-Pentyne, Sigma-Aldrich®
Catalog # SIAL271357-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:68.12 g/mol
Boiling Pt:56 °C (1013 hPa)
Melting Pt:–109 °C
Density:0.7122 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:210-989-2
UN:1993
ADR:3,II
:M-Clarity™ quality level = MQ100
CAS Number:627-21-4
Formula:C₅H₈
MDL Number:MFCD00009380