4,4-Dimethyl-1-pentene, Sigma-Aldrich®
SIAL110701-5G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
:
:98.19 g/mol
:73
:–136.6 °C
:0.6827 g/cm³ (20 °C)
:Ambient
:212-101-9
:3295
:762-62-9
:C₇H₁₄
:MFCD00026337
:3 II
:M-Clarity™ quality level = MQ100