3-Methyl-1-butene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:563-45-1
:
Formula:C₅H₁₀
MDL Number:MFCD00008937
:70.13 g/mol
Boiling Pt:20 °C (1013 hPa)
Melting Pt:–168 °C
Density:0.6272 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:209-249-1
ADR:3,I
:M-Clarity™ quality level = MQ100
UN:2561
Frequently Bought Together