Maleic acid ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110-16-7
:
Formula:C₄H₄O₄
MDL Number:MFCD00063177
:116.07 g/mol
Merck Index:14,5703
Boiling Pt:256 °C (1013 hPa)
Melting Pt:132…136 °C
Density:1.59
Flash Pt:127 °C (260 °F)
Storage Temperature:Ambient
EINECS:203-742-5
UN:3261
ADR:8,III
:GHS07
Related Information
Frequently Bought Together