HEPES (2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulphonic acid), Sigma-Aldrich®
: Merck
:7365-45-9
:
:C₈H₁₈N₂O₄S
:MFCD00006158
:13,04672
:210…215 °C
:1.448 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ400
:238.31 g/mol
:Ambient
:230-907-9
Frequently Bought Together