
2,5-Dimethyl-2,5-hexanediol ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:110-03-2
:
Formula:C₈H₁₈O₂
MDL Number:MFCD00004473
:146.23 g/mol
Boiling Pt:214…215 °C
Melting Pt:87…91 °C
Density:0.898
Flash Pt:126 °C (258 °F)
EINECS:203-731-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together