Order Entry
Slovenia
ContactUsLinkComponent
Ammonia, anhydrous ≥99.95%, puriss., Sigma-Aldrich®
Compressed Gas
Corrosive
Toxic
Environment
Ammonia, anhydrous ≥99.95%, puriss., Sigma-Aldrich®
Catalog # SIAL09682-1KG
Supplier:  Merck
CAS Number:  
Compressed Gas
Corrosive
Toxic
Environment
Ammonia, anhydrous ≥99.95%, puriss., Sigma-Aldrich®
Catalog # SIAL09682-1KG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Cylinder
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Vapor Density:
    0.6 (vs Air)
  • Vapor Pressure:
    8.75 atm (21 °C)
  • Assay:
    ≥99.95%
  • Autoignition Temperature:
    1203 °F
  • Explosion Limit:
    0.25
  • Impurities:
    ≤0.01% water

Specification Test Results

About this item

 :
 :17.03 g/mol
Boiling Pt:–33 °C (lit.)
Melting Pt:–78 °C (lit.)
Flash Pt:132 °C
EINECS:231-635-3
UN:1005
ADR:2.3(8),
 :GHS04
 :GHS05
 :GHS06
 :GHS09
CAS Number:7664-41-7
Formula:NH₃
MDL Number:MFCD00011418