Order Entry
Slovenia
Orders LinkContactUsLinkComponent
2-Ethylhexanal, Sigma-Aldrich®

2-Ethylhexanal, Sigma-Aldrich®

Supplier:   Merck
2-Ethylhexanal, Sigma-Aldrich®
Supplier:  Merck
Certificates
2-Ethylhexanal, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
  • CAS Number:123-05-7
  • Formula:C₈H₁₆O
  • MDL Number:MFCD00006987
  • Molecular Weight:128.21 g/mol
  • Boiling Pt:163 °C (1013 hPa)
  • Melting Pt:–67 °C
  • Density:0.82 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:204-596-5
  • UN:1989
  • ADR:3,III
  •  :M-Clarity™ quality level = MQ100
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    5 mL
    Log in for availability
    Log in for availability
    1 * 5 mL
    1 * 5 mL - 46,20 €
    46,20 €
    1 * 5 mL
       46,20 €

  • Glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 111,00 €
    111,00 €
    1 * 250 mL
       111,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.