Order Entry
Slovenia
ContactUsLinkComponent
Methanesulphonic acid ≥98%, TECHNICAL
Irritant
Corrosive
Methanesulphonic acid ≥98%, TECHNICAL
Supplier:  VWR Chemicals
Certificates
undefined
Irritant
Corrosive
Methanesulphonic acid ≥98%, TECHNICAL
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 98 %

Specification Test Results

Related Information

Frequently Bought Together