1-Hexadecanethiol, Sigma-Aldrich®
Catalog # SIAL674516-500MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:258.51 g/mol
Boiling Pt:335 °C (1013 hPa)
Melting Pt:18…20 °C
Density:0.8556 g/cm³ (20 °C)
Flash Pt:99 °C
Storage Temperature:Refrigerator
EINECS:220-846-6
UN:3334
ADR:9,
:M-Clarity™ quality level = MQ100
CAS Number:2917-26-2
Formula:C₁₆H₃₄S
MDL Number:MFCD00011677