Order Entry
Slovenia
ContactUsLinkComponent
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol), Sigma-Aldrich®
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol), Sigma-Aldrich®
  ROCH11814273001
 :  ROCHE
 :  
undefined
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol), Sigma-Aldrich®
  ROCH11814273001
 :  ROCHE
 :  
  • Pack type:
    Plastic bottle for solids
  • Pk:
    5 kg
  • Seq:
    4114
 :
 :121.14 g/mol
 :13,09842
 :357 °C (1013 hPa)
 :169…172 °C
 :1.328 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
 :Ambient
 :201-064-4
 :GHS07
 :77-86-1
 :NH₂C(CH₂OH)₃
 :MFCD00004679