
Citric acid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:77-92-9
:
Formula:HOC(COOH)(CH₂COOH)₂
MDL Number:MFCD00011669
:192.13 g/mol
Merck Index:13,02350
Boiling Pt:310 °C (1013 hPa)
Melting Pt:149…151 °C
Density:1.665 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:201-069-1
REACH:01-2119457026-42-0000
:GHS07
Frequently Bought Together