L-Methionine
Supplier: BIOMOL RESEARCH LABORATORIES
Certificates
About this item
CAS Number:63-68-3
:
Formula:C₅H₁₁NO₂S
MDL Number:MFCD00063097
:149.21 g/mol
Merck Index:13,06004
Boiling Pt:278 °C (1013 hPa)
Melting Pt:136 °C
Density:1.373 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-562-9
Frequently Bought Together