Order Entry
Slovenia
ContactUsLinkComponent
Glycine
Glycine
Catalog # EHERC14037000
Supplier:  EHRENSTORFER
CAS Number:  
undefined
Glycine
Catalog # EHERC14037000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :75.07 g/mol
Merck Index:13,04504
Boiling Pt:181 °C (1013 hPa)
Melting Pt:83 °C
Density:1.64 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-272-2
CAS Number:56-40-6
Formula:NH₂CH₂COOH
MDL Number:MFCD00008131