Order Entry
Slovenia
Orders LinkContactUsLinkComponent
2-Naphthyl acetate ≥99%

2-Naphthyl acetate ≥99%

2-Naphthyl acetate ≥99%
Certificates
2-Naphthyl acetate ≥99%
2-Naphthyl acetate ≥99%

3 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 1523-11-1 Molecular Formula: C12H10O2 Formula Weight: 186.21 Storage Temperature: 2°C to 8°C Melting Point: 68-70° Beilstein Reference: 1868291 MDL No.: MFCD00004060

Synonyms:  Acetic acid β-naphthyl ester, Ethanoic acid β-naphthyl ester, Acetic acid 2-naphthyl ester, β-Naphthyl acetate, Ethanoic acid 2-naphthyl ester
  • CAS Number:1523-11-1
  • Formula:C₁₂H₁₀O₂
  • MDL Number:MFCD00004060
  • Molecular Weight:186.21 g/mol
  • Boiling Pt:310 °C (1013 hPa)
  • Melting Pt:68…70 °C
  • Density:1.1108 g/cm³ (70 °C)
  • Storage Temperature:Refrigerator
  • EINECS:216-194-7
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.