Order Entry
Slovenia
Orders LinkContactUsLinkComponent
1-Naphthyl butyrate 98%

1-Naphthyl butyrate 98%

1-Naphthyl butyrate 98%
Certificates
1-Naphthyl butyrate 98%

2 Options Available Below

Product Details & Documents
Synonyms:  Butyric acid 1-naphthyl ester, α-Naphthyl butyrate, Butanoic acid α-naphthyl ester, Butanoic acid 1-naphthyl ester, Butyric acid α-naphthyl ester
  • CAS Number:3121-70-8
  • Formula:C₁₄H₁₄O₂
  • MDL Number:MFCD00003923
  • Molecular Weight:214.26 g/mol
  • Boiling Pt:125…130 °C (1 mmHg)
  • Melting Pt:95,5 - 96.5 °C
  • Density:1.09 g/cm³
  • Flash Pt:62 °C
  • Storage Temperature:Ambient
  • EINECS:221-500-7

Documents

Certificates
Search by Lot Number
Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear yellow Liquid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥97.5 %
Product Family Options

Product Information

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.