Order Entry
Slovenia
ContactUsLinkComponent
 
Methyl octanoate
Catalog # EHERC15711090
Supplier:  EHRENSTORFER
CAS Number:  
Methyl octanoate
Catalog # EHERC15711090
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Caprylic acid methyl ester, Octanoic acid methyl ester, Methyl caprylate
 :
 :158.24 g/mol
EINECS:203-835-0
CAS Number:111-11-5
Formula:C₉H₁₈O₂
MDL Number:MFCD00009551
Boiling Pt:190
Melting Pt:–36.7 °C
Density:0.877 g/cm³ (20 °C)
Flash Pt:68