Order Entry
Slovenia
ContactUsLinkComponent
 
Flammable
Irritant
1-Bromo-2-propanol
Catalog # EHERC10760000
Supplier:  EHRENSTORFER
CAS Number:  
Flammable
Irritant
1-Bromo-2-propanol
Catalog # EHERC10760000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :138.99 g/mol
Boiling Pt:145 °C (1013 hPa)
Melting Pt:–55 °C
Density:1.53 g/cm³ (20 °C)
Flash Pt:54 °C
Storage Temperature:Ambient
EINECS:243-225-1
UN:1993
ADR:3,III
 :GHS02
 :GHS07
CAS Number:19686-73-8
Formula:BrH₂CCH(OH)CH₃
MDL Number:MFCD00004529