2-Bromopropane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:75-26-3
:
Formula:H₃CBrCHCH₃
MDL Number:MFCD00000147
Merck Index:13,05230
Boiling Pt:59 °C (1013 hPa)
Melting Pt:–89 °C
Density:1.31 g/cm³ (20 °C)
Flash Pt:1 °C
EINECS:200-855-1
UN:2344
ADR:3,II
:M-Clarity™ quality level = MQ200
:122.99 g/mol
:GHS02
:GHS08
Storage Temperature:Ambient
Frequently Bought Together