D-(-)-Mannitol, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
- CAS Number:69-65-8
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00064287
- Molecular Weight:182.17 g/mol
- Merck Index:13,05769
- Boiling Pt:290…295 °C (4 hPa)
- Melting Pt:164…169 °C
- Density:1.49 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ200
- Storage Temperature:Ambient
- EINECS:200-711-8
5 Options Available Below
Product Details & Documents
Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
- CAS Number:69-65-8
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00064287
- Molecular Weight:182.17 g/mol
- Merck Index:13,05769
- Boiling Pt:290…295 °C (4 hPa)
- Melting Pt:164…169 °C
- Density:1.49 g/cm³ (20 °C)
- :M-Clarity™ quality level = MQ200
- Storage Temperature:Ambient
- EINECS:200-711-8
Certificates
Search by Lot NumberProduct Family Options
Product Information
- Pack typePkAvailabilityPrice
This product has been blocked by your organization. Please contact your purchasing department for more information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



