Order Entry
Slovenia
ContactUsLinkComponent
 
Organic reference standard, 2,3,3',4,5-Pentachlorobiphenyl (PCB No. 106) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20010600IO
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, 2,3,3',4,5-Pentachlorobiphenyl (PCB No. 106) 10 µg/ml in isooctane, Dr Ehrenstorfer
Catalog # EHERL20010600IO
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    70424-69-0
  • Description:
    2,3,3',4,5-Pentachlorobiphenyl (PCB No. 106) 10 µg/ml in isooctane
  • Formula:
    C₁₂H₅Cl₅
  • Description:
    2,3,3',4,5-Pentachlorobiphenyl (PCB No. 106)

Specifications

About this item