L-Arginine hydrochloride
Supplier: Roth Carl
Certificates
About this item
CAS Number:1119-34-2
:
Formula:C₆H₁₅ClN₄O₂
MDL Number:MFCD00064550
:210.66 g/mol
Merck Index:13,00785
Melting Pt:218 °C
Density:1.42 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:214-275-1
Frequently Bought Together