L(+)-Asparagine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:70-47-3
:
Formula:H₂NCOCH₂CH(NH₂)CO₂H
MDL Number:MFCD00064401
:132.12 g/mol
Merck Index:12,00872
Boiling Pt:343 °C (1013 hPa)
Melting Pt:234…235 °C
:M-Clarity™ quality level = MQ400
Storage Temperature:Ambient
EINECS:200-735-9
Frequently Bought Together