(S)-(-)-2-Bromopropionic acid, Sigma-Aldrich®
Catalog # SIAL385514-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:152.98 g/mol
Boiling Pt:203 °C (1013 hPa)
Melting Pt:–0.2 °C
Density:1.7 g/cm³ (20 °C)
Storage Temperature:Refrigerator
UN:3265
ADR:8,II
:M-Clarity™ quality level = MQ200
CAS Number:32644-15-8
Formula:H₃CBrCHCOOH
MDL Number:MFCD00137325