Organic reference standard, Chloroethane 2000 µg/ml in methanol
Catalog # REST30263-3
Supplier: Restek
CAS Number:
Specifications
- Pack type:Glass ampoule
- Pk:1 Ampoul
- CAS No.:75-00-3
- Description:Chloroethane 2,000 µg/ml in methanol
- Formula:C₂H₅Cl
- Description:Chloroethane