Organic reference standard, 1,2-Dichloroethane 10 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERL12422200ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:107-06-2
- Description:1,2-Dichloroethane 10 µg/ml in methanol
- Environmentally Preferable:
- Formula:C₂H₄Cl₂
- Description:1,2-Dichloroethane