Organic reference standard, 1,1,1-Trichloroethane 10 µg/ml in methanol, Dr Ehrenstorfer
EHERL17738300ME
:
- Pack type:Glass bottle
- Pk:10 mL
- CAS No.:71-55-6
- Description:1,1,1-Trichloroethane 10 µg/ml in methanol
- Formula:C₂H₃Cl₃
- Description:1,1,1-Trichloroethane