
Chromium(III) chloride hexahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10060-12-5
:
Formula:CrCl₃·6H₂O
MDL Number:MFCD00149660
:266.45 g/mol
Merck Index:13,02242
Melting Pt:85 °C
Density:1.76 g/cm³ (20 °C)
EINECS:233-038-3
ADR:8,III
:M-Clarity™ quality level = MQ200
UN:3260
:GHS07
Storage Temperature:Ambient
Frequently Bought Together