ESD safety shoes, sandals, uvex 1 ladies, 8562
: UVEX
Metal-free, extremely lightweight, flexible and multifunctional safety shoe made from breathable water resistant high-tech micro velour upper, soft padded tongue and collar, and a dual-density PUR sole (oil and petrol resistant, anti-static) free of lacquer wetting inhibitors such as silicones or phthalates.
- Climazone technology for improved comfort
- Breathable distance mesh lining
- Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
- Anti-slip sole with shock absorption at heel and front of the foot
- Virtually seamless design to eliminate the potential for pressure points and blisters
- 100% metal-free xenova® toecap, non magnetic (200 J)
- Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
Suitable for people allergic to chrome.
Colour: Grey/pink
Width: 10
: EN 61340 (ESD); DGUV112-191; EN ISO 20345:2011; S1, SRC
Frequently Bought Together