


Copper(II) nitrate hemipentahydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:19004-19-4
:
Formula:Cu(NO₃)₂·2.5H₂O
MDL Number:MFCD02253001
:232.59 g/mol
Melting Pt:255 °C
Density:>1 g/cm³ (20 °C)
EINECS:221-838-5
UN:1477
ADR:5.1,II
:M-Clarity™ quality level = MQ200
:GHS03
:GHS07
:GHS09
Frequently Bought Together