
Chromium(III) potassium sulphate dodecahydrate
Supplier: Roth Carl
Certificates
About this item
CAS Number:7788-99-0
:
Formula:CrK(SO₄)₂·12H₂O
MDL Number:MFCD00149917
:499.4 g/mol
Merck Index:13,02250
Melting Pt:89 °C
Density:1.83 g/cm³ (20 °C)
EINECS:233-401-6
:GHS07
Frequently Bought Together